3-(2-Aminoethyl)thiophene hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Thiophen-3-yl)ethanamine hydrochloride, 2-(Thien-3-yl)ethylamine hydrochloride

OR40114-1G
APOSOR40114-1GEA 62.5 EUR
APOSOR40114-1G
3-(2-Aminoethyl)thiophene hydrochloride
3-(2-Aminoethyl)thiophene hydrochloride
Formel: C₆H₉NS·ClH
Molecular Weight: 163,668 g/mol
Schmelzpunkt: 214…216 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09265521
CAS-Nummer: 34843-84-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR