5-Bromo-1H-benzoimidazole-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR40115-250MG
APOSOR40115-250MGEA 0 EUR
APOSOR40115-250MG
5-Bromo-1H-benzoimidazole-2-carboxylic acid
5-Bromo-1H-benzoimidazole-2-carboxylic acid
Formel: C₈H₅BrN₂O₂
Schmelzpunkt: 158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06738769
CAS-Nummer: 40197-20-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR