8-Methoxyquinoline-5-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Carboxy-8-methoxyquinoline

OR40116-250MG
APOSOR40116-250MGEA 300 EUR
APOSOR40116-250MG
8-Methoxyquinoline-5-carboxylic acid 95%
8-Methoxyquinoline-5-carboxylic acid
Formel: C₁₁H₉NO₃
Molecular Weight: 203,194 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08275136
CAS-Nummer: 148887-46-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR