4-Bromo-6-methyl-1H-indazole

Lieferant: Apollo Scientific
Achtung

OR40119-250MG
APOSOR40119-250MGEA 112 EUR
APOSOR40119-250MG
4-Bromo-6-methyl-1H-indazole
4-Bromo-6-methyl-1H-indazole
Formel: C₈H₇BrN₂
Molecular Weight: 211,059 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07781508
CAS-Nummer: 885521-94-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR