3-(4-Morpholinylcarbonyl)phenylboronsäure 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-(morpholine-4-carbonyl)phenyl)boronic acid, 3-(Morpholine-4-carbonyl)phenylboronic acid, 3-(4-Morpholinylcarbonyl)benzeneboronic acid

OR4012-1G
APOSOR4012-1GEA 949 EUR
APOSOR4012-1G
3-(4-Morpholinylcarbonyl)phenylboronsäure 96%
3-(4-Morpholinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₄BNO₄
Molecular Weight: 235,05 g/mol
Schmelzpunkt: 124…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115687
CAS-Nummer: 723281-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR