4-Bromo-1,5-dimethyl-1H-indazole

Lieferant: Apollo Scientific
Achtung

OR40124-250MG
APOSOR40124-250MGEA 21.9 EUR
APOSOR40124-250MG
4-Bromo-1,5-dimethyl-1H-indazole
4-Bromo-1,5-dimethyl-1H-indazole
Formel: C₉H₉BrN₂
Lagertemperatur: Kühlschrank
MDL: MFCD12028620
CAS-Nummer: 1159511-77-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR