5-Bromo-1,3-dimethyl-1H-indazole 98%

Lieferant: Apollo Scientific

OR40127-250MG
APOSOR40127-250MGEA 71.2 EUR
APOSOR40127-250MG
5-Bromo-1,3-dimethyl-1H-indazole 98%
5-Bromo-1,3-dimethyl-1H-indazole
Formel: C₉H₉BrN₂
Molecular Weight: 225,085 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12028623
CAS-Nummer: 552331-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR