5-Bromo-2,4-dimethyl-2H-indazole

Lieferant: Apollo Scientific
Achtung

OR40139-250MG
APOSOR40139-250MGEA 27.7 EUR
APOSOR40139-250MG
5-Bromo-2,4-dimethyl-2H-indazole
5-Bromo-2,4-dimethyl-2H-indazole
Formel: C₉H₉BrN₂
Lagertemperatur: Kühlschrank
MDL: MFCD12028635
CAS-Nummer: 1159511-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR