5-Bromo-2,7-dimethyl-2H-indazole 95%

Lieferant: Apollo Scientific
Achtung

OR40141-250MG
APOSOR40141-250MGEA 43.5 EUR
APOSOR40141-250MG
5-Bromo-2,7-dimethyl-2H-indazole 95%
5-Bromo-2,7-dimethyl-2H-indazole
Formel: C₉H₉BrN₂
Lagertemperatur: Kühlschrank
MDL: MFCD12028637
CAS-Nummer: 1146637-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR