5-Methyl-1H-indazol-4-boronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(5-Methyl-1H-indazol-4-yl)boronic acid
OR40147-100MG
APOSOR40147-100MGEA
21.9
EUR
APOSOR40147-100MG
5-Methyl-1H-indazol-4-boronsäure 97%
5-Methyl-1H-indazole-4-boronic acid
Formel:
C₈H₉BN₂O₂ Molecular Weight: 175,9800 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10001490 CAS-Nummer: 1245816-10-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...