3-Methyl-1H-indazol-5-boronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Methyl-1H-indazol-5-yl-5-boronic acid

OR40150-250MG
APOSOR40150-250MGEA 46.4 EUR
APOSOR40150-250MG
3-Methyl-1H-indazol-5-boronsäure
3-Methyl-1H-indazole-5-boronic acid
Formel: C₈H₉BN₂O₂
Molecular Weight: 175,9800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12028643
CAS-Nummer: 1245816-25-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR