4-Methyl-1H-indazol-5-boronsäure 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Methyl-1H-indazol-5-yl)boronic acid

OR40151-500MG OR40151-1G OR40151-5G
APOSOR40151-500MGEA 39.3 EUR
APOSOR40151-500MG APOSOR40151-1G APOSOR40151-5G
4-Methyl-1H-indazol-5-boronsäure 96%
4-Methyl-1H-indazole-5-boronic acid
Formel: C₈H₉BN₂O₂
Molecular Weight: 175.9800 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12028644
CAS-Nummer: 1245816-09-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR