6-Methyl-1H-indazole-5-boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (6-Methyl-1H-indazol-5-yl)boronic acid, 5-Borono-6-methyl-1H-indazole

OR40152-250MG
APOSOR40152-250MGEA 60.9 EUR
APOSOR40152-250MG
6-Methyl-1H-indazole-5-boronic acid
6-Methyl-1H-indazole-5-boronic acid
Formel: C₈H₉BN₂O₂
Schmelzpunkt: 115…119 °C
Lagertemperatur: Kühlschrank
MDL: MFCD12028645
CAS-Nummer: 1310405-35-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR