1,3-Dimethyl-1H-indazole-5-boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Borono-1,3-dimethyl-1H-indazole, (1,3-Dimethyl-1H-indazol-5-yl)boronic acid

OR40161-25MG
APOSOR40161-25MGEA 24.8 EUR
APOSOR40161-25MG
1,3-Dimethyl-1H-indazole-5-boronic acid
1,3-Dimethyl-1H-indazole-5-boronic acid
Formel: C₉H₁₁BN₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD12028654
CAS-Nummer: 1310404-48-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR