3,4-Dimethoxybenzophenon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3,4-Dimethoxyphenyl)(phenyl)methanone

OR40183-100G
APOSOR40183-100GEA 81.2 EUR
APOSOR40183-100G
3,4-Dimethoxybenzophenon 98%
3,4-Dimethoxybenzophenon
Formel: C₁₅H₁₄O₃
Molecular Weight: 242.2700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00075875
CAS-Nummer: 4038-14-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR