2,4-Dichlor-5-iodpyrimidin

Lieferant: Apollo Scientific
Gefahr

OR40189-1G
APOSOR40189-1GEA 21.9 EUR
APOSOR40189-1G
2,4-Dichlor-5-iodpyrimidin
2,4-Dichloro-5-iodopyrimidine
Formel: C₄HCl₂IN₂
Molecular Weight: 274,87 g/mol
Schmelzpunkt: 65…69 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01898087
CAS-Nummer: 13544-44-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR