(3,5-Dimethylisoxazol-4-yl)methylamine
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(3,5-Dimethylisoxazol-4-yl)methanamine
OR40198-1G
APOSOR40198-1GEA
223
EUR
APOSOR40198-1G
(3,5-Dimethylisoxazol-4-yl)methylamine
(3,5-Dimethylisoxazol-4-yl)methylamine
Formel:
C₆H₁₀N₂O Molecular Weight: 126,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06213033 CAS-Nummer: 131052-47-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...