(R)-2-Benzyl-3-((tert-butoxycarbonyl)amino)propanoic acid

Lieferant: Apollo Scientific

OR40214-100MG
APOSOR40214-100MGEA 0 EUR
APOSOR40214-100MG
(R)-2-Benzyl-3-((tert-butoxycarbonyl)amino)propanoic acid
(R)-2-Benzyl-3-((tert-butoxycarbonyl)amino)propanoic acid
Formel: C₁₅H₂₁NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD07372496
CAS-Nummer: 262301-38-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR