7-Isochinolinboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: Isoquinolin-7-ylboronic acid

OR40225-250MG
APOSOR40225-250MGEA 66.7 EUR
APOSOR40225-250MG
7-Isochinolinboronsäure
7-Isoquinolineboronic acid
Formel: C₉H₈BNO₂
Molecular Weight: 172.9763 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08234617
CAS-Nummer: 1092790-21-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR