1H-Indazole-7-carboxaldehyde 95%

Lieferant: Apollo Scientific
Achtung

OR40230-1G
APOSOR40230-1GEA 59.6 EUR
APOSOR40230-1G
1H-Indazole-7-carboxaldehyde 95%
1H-Indazole-7-carboxaldehyde
Formel: C₈H₆N₂O
Lagertemperatur: Kühlschrank
MDL: MFCD06738281
CAS-Nummer: 312746-72-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR