2-(Bromomethyl)-4,6-dimethylpyridine 95%

Lieferant: Apollo Scientific
Achtung

OR40233-100MG
APOSOR40233-100MGEA 0 EUR
APOSOR40233-100MG
2-(Bromomethyl)-4,6-dimethylpyridine 95%
2-(Bromomethyl)-4,6-dimethylpyridine
Formel: C₈H₁₀BrN
Lagertemperatur: Kühlschrank
MDL: MFCD10699165
CAS-Nummer: 79313-01-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR