5-Chloro-3-methyl-1H-indole 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR40238-100MG
APOSOR40238-100MGEA
0
EUR
APOSOR40238-100MG
5-Chloro-3-methyl-1H-indole 97%
5-Chloro-3-methyl-1H-indole
Formel:
C₉H₈ClN Schmelzpunkt: 72 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD04972055 CAS-Nummer: 71095-42-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...