2-Methyl-4-hydroxymethylpyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Methylpyridin-4-yl)methanol

OR40250-1G
APOSOR40250-1GEA 144 EUR
APOSOR40250-1G
2-Methyl-4-hydroxymethylpyridine
2-Methyl-4-hydroxymethylpyridine
Formel: C₇H₉NO
Molecular Weight: 123,16 g/mol
Schmelzpunkt: 58…59 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08062415
CAS-Nummer: 105250-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR