Isochinolin-3-carbaldehyd 95%

Lieferant: Apollo Scientific
Achtung

OR40275-250MG
APOSOR40275-250MGEA 31.9 EUR
APOSOR40275-250MG
Isochinolin-3-carbaldehyd 95%
Isochinolin-3-carbaldehyd
Formel: C₁₀H₇NO
Molecular Weight: 157,1700 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06227437
CAS-Nummer: 5470-80-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR