Methyl-2-bromo-5-chlorothiazole-4-carboxylate 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR40279-250MG
APOSOR40279-250MGEA
88.6
EUR
APOSOR40279-250MG
Methyl-2-bromo-5-chlorothiazole-4-carboxylate 98%
Methyl-2-bromo-5-chlorothiazole-4-carboxylate
Formel:
C₅H₃BrClNO₂S Lagertemperatur: Kühlschrank |
MDL:
MFCD08704624 CAS-Nummer: 1053655-63-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...