N-tert-Butyl-3-boronobenzolsulfonamid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-[N-(tert-Butyl)sulphamoyl]phenylboronic Acid

OR4028-250MG
APOSOR4028-250MGEA 52.2 EUR
APOSOR4028-250MG
N-tert-Butyl-3-boronobenzolsulfonamid 97%
N-tert-Butyl-3-boronobenzolsulfonamid
Formel: C₁₀H₁₆BNO₄S
Molecular Weight: 257,1143 g/mol
Schmelzpunkt: 120…124 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06659842
CAS-Nummer: 221290-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR