[5-(2-Methyl-1,3-thiazol-4-yl)thien-2-yl]methylamine dihydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Aminomethyl)-5-(2-methyl-1,3-thiazol-4-yl)thiophene dihydrochloride, 4-[5-(Aminomethyl)thien-2-yl]-2-methyl-1,3-thiazole dihydrochloride
OR40290-100MG
APOSOR40290-100MGEA
151
EUR
APOSOR40290-100MG
[5-(2-Methyl-1,3-thiazol-4-yl)thien-2-yl]methylamine dihydrochloride
[5-(2-Methyl-1,3-thiazol-4-yl)thien-2-yl]methylamine dihydrochloride
Formel:
C₉H₁₀N₂S₂·2ClH Molecular Weight: 283,241 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD07704030 CAS-Nummer: 1030012-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...