N-tert-Butyl-4-boronobenzolsulfonamid 97%

Lieferant: Apollo Scientific
Achtung

OR4030-1G OR4030-250MG OR4030-5G
APOSOR4030-1GEA 120 EUR
APOSOR4030-1G APOSOR4030-250MG APOSOR4030-5G
N-tert-Butyl-4-boronobenzolsulfonamid 97%
N-tert-Butyl-4-boronobenzolsulfonamid
Formel: C₁₀H₁₆BNO₄S
Schmelzpunkt: 128…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659841
CAS-Nummer: 208516-15-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR