Methyl-2-amino-5-methylthiazole-4-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-amino-5-methylthiazole-4-carboxylate

OR40302-1G
APOSOR40302-1GEA 106 EUR
APOSOR40302-1G
Methyl-2-amino-5-methylthiazole-4-carboxylate
Methyl-2-amino-5-methylthiazole-4-carboxylate
Formel: C₆H₈N₂O₂S
Molecular Weight: 172.21 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04035351
CAS-Nummer: 63257-03-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR