trans-Diethyl tetrahydro-1H-pyrrole-3,4-dicarboxylate 95+%
Lieferant: Apollo Scientific
Synonyme:
Diethyl trans-pyrrolidine-3,4-dicarboxylate
OR40307-500MG
APOSOR40307-500MGEA
560
EUR
APOSOR40307-500MG
trans-Diethyl tetrahydro-1H-pyrrole-3,4-dicarboxylate 95+%
trans-Diethyl tetrahydro-1H-pyrrole-3,4-dicarboxylate
Formel:
C₁₀H₁₇NO₄ Molecular Weight: 215,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16140346 CAS-Nummer: 904304-88-7 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...