3-[Di(but-1-yl)amino]propan-1-ol
Lieferant: Apollo Scientific
|
Achtung
|
OR40320-1G
APOSOR40320-1GEA
34.8
EUR
APOSOR40320-1G
3-[Di(but-1-yl)amino]propan-1-ol
3-[Di(but-1-yl)amino]propan-1-ol
Formel:
C₁₁H₂₅NO Siedepunkt: 94…97 °C (3 mm Hg) Lagertemperatur: Kühlschrank |
MDL:
MFCD00152393 CAS-Nummer: 2050-51-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...