4-Cyclohexylthiophenol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Cyclohexylbenzenethiol, 4-Cyclohexylphenyl mercaptan

OR40333-250MG
APOSOR40333-250MGEA 557 EUR
APOSOR40333-250MG
4-Cyclohexylthiophenol
4-Cyclohexylthiophenol
Formel: C₁₂H₁₆S
Molecular Weight: 192,32 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00068897
CAS-Nummer: 18325-50-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR