3-Amino-2-brom-5-methylpyridin 98%
Lieferant: Apollo Scientific
Synonyme:
3-Amino-2-bromo-5-picoline, 5-Amino-6-bromo-3-picoline, 2-Bromo-5-methyl-3-pyridinamine
OR40336-5G
APOSOR40336-5GEA
66.7
EUR
APOSOR40336-5G
3-Amino-2-brom-5-methylpyridin 98%
3-Amino-2-bromo-5-methylpyridine
Formel:
C₆H₇BrN₂ Schmelzpunkt: 103…106 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08277281 CAS-Nummer: 34552-14-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...