(2-Chloropyridin-3-yl)acetic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR40337-250MG
APOSOR40337-250MGEA
0
EUR
APOSOR40337-250MG
(2-Chloropyridin-3-yl)acetic acid 97%
(2-Chloropyridin-3-yl)acetic acid
Formel:
C₇H₆ClNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09757512 CAS-Nummer: 61494-55-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...