(2-Chloropyridin-3-yl)acetonitrile 97%

Lieferant: Apollo Scientific
Gefahr

OR40338-10G OR40338-25G OR40338-5G
APOSOR40338-10GEA 173 EUR
APOSOR40338-10G APOSOR40338-25G APOSOR40338-5G
(2-Chloropyridin-3-yl)acetonitrile 97%
(2-Chloropyridin-3-yl)acetonitrile
Formel: C₇H₅ClN₂
Molecular Weight: 152.5800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09923767
CAS-Nummer: 101012-32-2
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR