5-Chloro-2-oxo-2,3-dihydro-1,3-benzoxazol-7-ylbenzenecarboxylate 95+%

Lieferant: Apollo Scientific

Synonyme: 5-Chloro-2,3-dihydro-2-oxo-1,3-benzoxazol-7-yl benzoate

OR40344-500MG
APOSOR40344-500MGEA 661 EUR
APOSOR40344-500MG
5-Chloro-2-oxo-2,3-dihydro-1,3-benzoxazol-7-ylbenzenecarboxylate 95+%
5-Chloro-2-oxo-2,3-dihydro-1,3-benzoxazol-7-ylbenzenecarboxylate
Formel: C₁₄H₈ClNO₄
Molecular Weight: 289,68 g/mol
Schmelzpunkt: 218…220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16622768
CAS-Nummer: 1228182-74-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR