3-Iodo-6-methoxypyridazine

Lieferant: Apollo Scientific

OR40348-1G
APOSOR40348-1GEA 333 EUR
APOSOR40348-1G
3-Iodo-6-methoxypyridazine
3-Iodo-6-methoxypyridazine
Formel: C₅H₅IN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD10697533
CAS-Nummer: 17321-35-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR