6-Chloro-7-iodo-7-deazapurine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine, 6-Chloro-7-iodo-deazapurine

OR40353-5G OR40353-1G OR40353-10G
APOSOR40353-5GEA 24.8 EUR
APOSOR40353-5G APOSOR40353-1G APOSOR40353-10G
6-Chloro-7-iodo-7-deazapurine 97%
6-Chloro-7-iodo-7-deazapurine
Formel: C₆H₃ClIN₃
Molecular Weight: 279.47 g/mol
Schmelzpunkt: 179…183 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD09263258
CAS-Nummer: 123148-78-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR