3,6-Dichloropyridine-2-carbonyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-(Chlorocarbonyl)-3,6-dichloropyridine, 3,6-Dichloropicolinoyl chloride

OR40358-1G
APOSOR40358-1GEA 102 EUR
APOSOR40358-1G
3,6-Dichloropyridine-2-carbonyl chloride
3,6-Dichloropyridine-2-carbonyl chloride
Formel: C₆H₂Cl₃NO
Molecular Weight: 210,445 g/mol
Siedepunkt: 100…104 °C (0,1 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD17676109
CAS-Nummer: 16866-53-8
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR