3-Chloro-6-methoxypyridine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-6-methoxypicolinic acid, 2-Carboxy-3-chloro-6-methoxypyridine

OR40360-1G
APOSOR40360-1GEA 132 EUR
APOSOR40360-1G
3-Chloro-6-methoxypyridine-2-carboxylic acid
3-Chloro-6-methoxypyridine-2-carboxylic acid
Formel: C₇H₆ClNO₃
Schmelzpunkt: 99…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11044291
CAS-Nummer: 856836-44-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR