3,6-Dichloropyridine-2-carboxamide
Lieferant: Apollo Scientific
|
Achtung
|
OR40361-1G
APOSOR40361-1GEA
227
EUR
APOSOR40361-1G
3,6-Dichloropyridine-2-carboxamide
3,6-Dichloropyridine-2-carboxamide
Formel:
C₆H₄Cl₂N₂O Schmelzpunkt: 178…180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00186645 CAS-Nummer: 1532-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...