2,5-Dichloropyridine-6-carboxaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,6-Dichloropicolinaldehyde
OR40362-250MG
APOSOR40362-250MGEA
21.9
EUR
APOSOR40362-250MG
2,5-Dichloropyridine-6-carboxaldehyde
2,5-Dichloropyridine-6-carboxaldehyde
Formel:
C₆H₃Cl₂NO Schmelzpunkt: 105…107 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12963768 CAS-Nummer: 343781-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...