3-Chloro-6-methoxypyridine-2-carbonyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Chloro-6-methoxypicolinoyl chloride, 3-Chloro-2-(chlorocarbonyl)-6-methoxypyridine

OR40363-1G
APOSOR40363-1GEA 467 EUR
APOSOR40363-1G
3-Chloro-6-methoxypyridine-2-carbonyl chloride
3-Chloro-6-methoxypyridine-2-carbonyl chloride
Formel: C₇H₅Cl₂NO₂
Molecular Weight: 206,026 g/mol
Schmelzpunkt: 42…44 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD17676110
CAS-Nummer: 1257535-51-5
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR