3,4-Dihydro-2-methyl-2H-1,4-benzoxazine

Lieferant: Apollo Scientific
Achtung

OR40370-1G
APOSOR40370-1GEA 232 EUR
APOSOR40370-1G
3,4-Dihydro-2-methyl-2H-1,4-benzoxazine
3,4-Dihydro-2-methyl-2H-1,4-benzoxazine
Formel: C₉H₁₁NO
Molecular Weight: 149.19 g/mol
Siedepunkt: 90 °C (1 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD07364074
CAS-Nummer: 58960-13-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR