7-Amino-2-methyl-2H-1,4-benzoxazin-3(4H)-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Amino-3,4-dihydro-2-methyl-3-oxo-2H-1,4-benzoxazine

OR40372-1G
APOSOR40372-1GEA 266 EUR
APOSOR40372-1G
7-Amino-2-methyl-2H-1,4-benzoxazin-3(4H)-one
7-Amino-2-methyl-2H-1,4-benzoxazin-3(4H)-one
Formel: C₉H₁₀N₂O₂
Molecular Weight: 178,188 g/mol
Schmelzpunkt: 173…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09882585
CAS-Nummer: 105807-79-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR