1-(5-Methyl-3-phenylisoxazol-4-yl)ethan-1-ol
Lieferant: Apollo Scientific
|
Achtung
|
OR40374-1G
APOSOR40374-1GEA
125
EUR
APOSOR40374-1G
1-(5-Methyl-3-phenylisoxazol-4-yl)ethan-1-ol
1-(5-Methyl-3-phenylisoxazol-4-yl)ethan-1-ol
Formel:
C₁₂H₁₃NO₂ Schmelzpunkt: 101…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD16631269 CAS-Nummer: 6497-24-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...