2-Amino-4-(thien-3-yl)-1,3-thiazole

Lieferant: Apollo Scientific

Synonyme: 4-(Thien-3-yl)-1,3-thiazol-2-amine

OR40375-1G
APOSOR40375-1GEA 422 EUR
APOSOR40375-1G
2-Amino-4-(thien-3-yl)-1,3-thiazole
2-Amino-4-(thien-3-yl)-1,3-thiazole
Formel: C₇H₆N₂S₂
Molecular Weight: 182,266 g/mol
Schmelzpunkt: 134…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD14707003
CAS-Nummer: 105362-05-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR