Benzyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate

Lieferant: Apollo Scientific

Synonyme: Benzyl 1-oxa-6-azaspiro[2.5]octane-6-carboxylate, 1-Oxa-6-azaspiro[2.5]octane, N-CBZ protected

OR40376-1G
APOSOR40376-1GEA 77 EUR
APOSOR40376-1G
Benzyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate
Benzyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate
Formel: C₁₄H₁₇NO₃
Molecular Weight: 247,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11976640
CAS-Nummer: 77211-75-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR