2-(Aminomethyl)-4-methylpyridine hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Methylpyridin-2-yl)methylamine hydrochloride, 4-Methylpicolinylamine hydrochloride

OR40440-1G OR40440-250MG
APOSOR40440-1GEA 288 EUR
APOSOR40440-1G APOSOR40440-250MG
2-(Aminomethyl)-4-methylpyridine hydrochloride 95%
2-(Aminomethyl)-4-methylpyridine hydrochloride
Formel: C₇H₁₀N₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD16628187
CAS-Nummer: 1257535-59-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR