2-(Chloromethyl)-1-methyl-1H-imidazole hydrochloride 95%

Lieferant: Apollo Scientific
Gefahr

OR40445-250MG
APOSOR40445-250MGEA 21.9 EUR
APOSOR40445-250MG
2-(Chloromethyl)-1-methyl-1H-imidazole hydrochloride 95%
2-(Chloromethyl)-1-methyl-1H-imidazole hydrochloride
Formel: C₅H₇ClN₂·ClH
Molecular Weight: 167.036 g/mol
Schmelzpunkt: 175…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00234118
CAS-Nummer: 78667-04-6
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR